Please login to your online account to display your discounted pricing

Mouse anti-Bcl-10, Clone: SPM520, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441501

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-10
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM520
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. O95999
Host Species Murine
Immunogen Human BCL10 recombinant protein (epitope aa122-168)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8915
Test Specificity This MAb labels subpopulations of normal B and T cells and is a useful tool for the sub-classification of lymphomas.
Reconstitution Protein A purified
Target Species Human