Please login to your online account to display your discounted pricing

Mouse anti-Bax, Clone: SPM336, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23280910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23280910

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Primate
Antigen Bax
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Host Species Mouse
Isotype IgG1
Quantity 1.0mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 581
Conjugate Unlabeled
Gene Accession No. Q07812
Immunogen A synthetic peptide
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Does not react to Mouse or Rat.
Clone SPM336
Concentration 0.2mg/mL