Please login to your online account to display your discounted pricing

Mouse anti-Bax, Clone: SPM336, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476301

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide
Isotype IgG1
Antigen Bax
Regulatory Status RUO
Quantity 0.1mg
Host Species Mouse
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Primate
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Test Specificity Recognizes a protein of 21kDa, identified as the Bax protein. This MAb is highly specific to Bax and shows no cross-reaction with Bcl-2a or Bcl-X protein. Does not react with Mouse or Rat.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Reconstitution Protein A purified
Format Purified
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q07812
Gene ID (Entrez) 581
Purification Method Purified
Clone SPM336
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.