Please login to your online account to display your discounted pricing

Mouse anti-Bax, Clone: SPM336, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476302

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Primate
Host Species Mouse
Gene Accession No. Q07812
Isotype IgG1
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Reconstitution Protein A purified
Quantity 0.2mg
Primary or Secondary Primary
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen A synthetic peptide
Format Purified
Gene ID (Entrez) 581
Antigen Bax
Conjugate Unlabeled
Test Specificity Recognizes a protein of 21kDa, identified as the Bax protein. This MAb is highly specific to Bax and shows no cross-reaction with Bcl-2a or Bcl-X protein. Does not react with Mouse or Rat.
Clone SPM336
Regulatory Status RUO
Purification Method Purified
Concentration 1mg/mL
Monoclonal or Polyclonal Monoclonal