Please login to your online account to display your discounted pricing

Mouse anti-B7-H4, Clone: H74, PE, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen B7-H4
Clone H74
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Purified B7H4.
Gene Accession No. Q7Z7D3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143743

 
novus biologicals canada
NBP143743100TESTS
100 Tests Each for $395.00

NBP14374325

 
novus biologicals canada
NBP14374325TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen B7-H4
Clone H74
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Purified B7H4.
Gene Accession No. Q7Z7D3
Gene ID (Entrez) 79679
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry