Please login to your online account to display your discounted pricing

Mouse anti-B7-H4, Clone: H74, Novus Biologicals™

$307.00 - $406.00

Specifications

Antigen B7-H4
Clone H74
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified B7H4.
Gene Accession No. Q7Z7D3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14327301

 
novus biologicals canada
NBP14327301MG
0.1mg Each for $406.00

NBP14327303

 
novus biologicals canada
NBP1432730025MG
0.025mg Each for $307.00
Description & Specifications

Specifications

Antigen B7-H4
Clone H74
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified B7H4.
Gene Accession No. Q7Z7D3
Gene ID (Entrez) 79679
Formulation PBS pH 7.2 may contain carrier protein/stabilizer.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry