Please login to your online account to display your discounted pricing

Mouse anti-B7-H4, Clone: H74, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1432730025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327303

  • / Each
 

Description & Specifications

Specifications

Antigen B7-H4
Applications Flow Cytometry
Clone H74
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS pH 7.2 may contain carrier protein/stabilizer.
Gene Accession No. Q7Z7D3
Host Species Murine
Immunogen The immunogen for this antibody was Purified B7H4.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 79679
Target Species Human