Please login to your online account to display your discounted pricing

Mouse anti-B7-H4, Clone: H74, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Format Affinity Purified
Gene ID (Entrez) 79679
Gene Accession No. Q7Z7D3
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Conjugate Unlabeled
Cross Reactivity Human
Primary or Secondary Primary
Antigen B7-H4
Clone H74
Purification Method Affinity Purified
Isotype IgG1, κ
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was Purified B7H4.
Host Species Mouse
Formulation PBS pH 7.2 may contain carrier protein/stabilizer.
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mg