Please login to your online account to display your discounted pricing

Mouse anti-B7-2/CD86, Clone: BU63, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23456901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23456901

  • / Each
 

Description & Specifications

Specifications

Format Purified
Gene Accession No. P42081
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen ARH-77 (B-lymphoblastoid cell line)
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Antigen B7-2/CD86
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 942
Reconstitution Protein A purified
Clone BU63
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat