Please login to your online account to display your discounted pricing

Mouse anti-B7-1/CD80, Clone: 3H5, PE, Novus Biologicals™

$252.00 - $362.00

Specifications

Antigen B7-1/CD80
Clone 3H5
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is CD80.
Gene ID (Entrez) 941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20043010

 
novus biologicals canada
NBP200430100UG
100μg Each for $252.00

NBP20043026

 
novus biologicals canada
NBP200430200UG
200μg Each for $362.00
Description & Specifications

Specifications

Antigen B7-1/CD80
Clone 3H5
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is CD80.
Gene ID (Entrez) 941
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry