Please login to your online account to display your discounted pricing

Mouse anti-B7-1/CD80, Clone: 3H5, Biotin, Novus Biologicals™

$252.00 - $428.00

Specifications

Antigen B7-1/CD80
Clone 3H5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14361801

 
novus biologicals canada
NBP14361801MG
0.1mg Each for $252.00

NBP1436185

 
novus biologicals canada
NBP14361805MG
0.5mg Each for $428.00
Description & Specifications

Specifications

Antigen B7-1/CD80
Clone 3H5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry