Please login to your online account to display your discounted pricing

Mouse anti-B7-1/CD80, Clone: 2D10.4, PE, Novus Biologicals™

$131.00 - $296.00

Specifications

Antigen B7-1/CD80
Clone 2D10.4
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143841

 
novus biologicals canada
NBP143841100TESTS
100 Tests Each for $296.00

NBP14384125

 
novus biologicals canada
NBP14384125TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen B7-1/CD80
Clone 2D10.4
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry