Please login to your online account to display your discounted pricing

Mouse anti-B7-1/CD80, Clone: 2D10.4, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen B7-1/CD80
Clone 2D10.4
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14338401

 
novus biologicals canada
NBP14338401MG
0.1mg Each for $285.00

NBP14338403

 
novus biologicals canada
NBP1433840025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen B7-1/CD80
Clone 2D10.4
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry