Please login to your online account to display your discounted pricing

Mouse anti-B7-1/CD80, Clone: 2D10.4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433840025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14338403

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Human
Regulatory Status RUO
Primary or Secondary Primary
Host Species Mouse
Gene ID (Entrez) 941
Antigen B7-1/CD80
Immunogen The immunogen for this antibody was CD80.
Isotype IgG1, κ
Purification Method Affinity Purified
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.025mg
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Clone 2D10.4
Conjugate Unlabeled