Please login to your online account to display your discounted pricing

Mouse anti-B7-1/CD80, Clone: 2D10.4, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen B7-1/CD80
Clone 2D10.4
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200431

 
novus biologicals canada
NBP200431100TESTS
100 Tests Each for $252.00

NBP20043125

 
novus biologicals canada
NBP20043125TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen B7-1/CD80
Clone 2D10.4
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay