Please login to your online account to display your discounted pricing

Mouse anti-Androgen R/NR3C4 (p Ser213, p Ser210), Clone: 156C135.2, Azide Free, Novus Biologicals™

$406.00 - $406.00

Specifications

Antigen Androgen R/NR3C4 (p Ser213, p Ser210)
Clone 156C135.2
Concentration 0.5mg/mL
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence.
Gene Accession No. P10275
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23109901

 
novus biologicals canada
NBP23109901MG
0.1mg Each for $406.00
Description & Specifications

Specifications

Antigen Androgen R/NR3C4 (p Ser213, p Ser210)
Clone 156C135.2
Concentration 0.5mg/mL
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence.
Gene Accession No. P10275
Gene ID (Entrez) 367
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting