Please login to your online account to display your discounted pricing

Mouse anti-Androgen R/NR3C4 (p Ser213, p Ser210), Clone: 156C135.2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23109901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23109901

  • / Each
 

Description & Specifications

Specifications

Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence.
Primary or Secondary Primary
Gene ID (Entrez) 367
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Format Purified
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.1mg
Isotype IgG1, κ
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Antigen Androgen R/NR3C4 (p Ser213, p Ser210)
Host Species Mouse
Formulation PBS. Protein G purified.
Conjugate Unlabeled
Gene Accession No. P10275
Concentration 0.5mg/mL
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Test Specificity In IGF-1 stimulated LNCaP cells (passage number 38), a ∽110kDa band was observed. Please see Lin et al. 2003 for additional details. The serine phosphorylation site recognized by this antibody has been alternatively referred to Ser213 (Lee and Chang, 2003) and Ser210 (Lin et al, 2003). Variations in denotation can arise from how the sequence is counted in various GenBank accession numbers. The site is denoted as Ser213 in GenBank Accession No. A39248, which was used to design the immunogen. Reacts with Human and New World Monkey. Predicted cross-reactivity with Chimpanzee, Dog, Mouse, Pig and Rat.
Clone 156C135.2