Please login to your online account to display your discounted pricing

Mouse anti-alpha-Fetoprotein/AFP, Clone: SPM334, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23291510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23291510

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 174
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Antigen alpha-Fetoprotein/AFP
Formulation PBS with 0.05% BSA.
Gene Accession No. P02771
Host Species Mouse
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity Does not react to Bovine, Feline, Mouse or Rat.
Reconstitution Protein A purified
Clone SPM334
Concentration 0.2mg/mL