Please login to your online account to display your discounted pricing

Mouse anti-alpha-Fetoprotein/AFP, Clone: SPM334, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23291501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23291501

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Format Purified
Gene Accession No. P02771
Host Species Mouse
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Isotype IgG2a, κ
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 174
Clone SPM334
Conjugate Unlabeled
Antigen alpha-Fetoprotein/AFP
Purification Method Purified
Regulatory Status RUO
Test Specificity Does not react to Bovine, Feline, Mouse or Rat.
Reconstitution Protein A purified
Concentration 0.2mg/mL