Please login to your online account to display your discounted pricing

Mouse anti-alpha-Fetoprotein/AFP, Clone: SPM334, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23775402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23775402

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Quantity 0.2mg
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Primary or Secondary Primary
Gene ID (Entrez) 174
Clone SPM334
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO
Concentration 1mg/mL
Purification Method Purified
Reconstitution Protein A purified
Test Specificity Antibody recognizes an oncofetal glycoprotein with a single chain of 70kDa, which is identified as alpha fetoprotein (AFP). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. Does not react with Bovine, Feline, Mouse or Rat.
Antigen alpha-Fetoprotein/AFP
Gene Accession No. P02771