Please login to your online account to display your discounted pricing

Mouse anti-alpha-Fetoprotein/AFP, Clone: C3, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23457701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23457701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Isotype IgG2a, κ
Antigen alpha-Fetoprotein/AFP
Host Species Mouse
Clone C3
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Conjugate Unlabeled
Gene Accession No. P02771
Gene ID (Entrez) 174
Quantity 0.1mg
Immunogen Alpha fetoprotein (AFP) purified from serum of a hepatoma patient
Primary or Secondary Primary
Format Purified
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Reconstitution Protein A purified
Test Specificity Does not react to Bovine, Feline, Mouse or Rat.