Please login to your online account to display your discounted pricing

Mouse anti-Agrin, Clone: Agr 33, Novus Biologicals™

$2,016.24 - $2,016.24

Specifications

Antigen Agrin
Clone Agr 33
Concentration 1.0mg/mL
Immunogen C-terminal construct.
Gene ID (Entrez) 375790
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NB120123581

 
novus biologicals canada
NB1201235810MG
1mg Each for $2,016.24
Description & Specifications

Specifications

Antigen Agrin
Clone Agr 33
Concentration 1.0mg/mL
Immunogen C-terminal construct.
Gene ID (Entrez) 375790
Formulation PBS, pH 7.2 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)