Please login to your online account to display your discounted pricing

Mouse anti-Agrin, Clone: Agr 33, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1201235810MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB120123581

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen C-terminal construct.
Isotype IgG1
Formulation PBS, pH 7.2 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Clone Agr 33
Concentration 1.0mg/mL
Format Purified
Quantity 1mg
Gene ID (Entrez) 375790
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Test Specificity Detects all isoforms of agrin. Cross-reacts with Mouse and Rat. Not yet tested in other species.
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Conjugate Unlabeled
Antigen Agrin
Purification Method Purified