Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 2 Smooth Muscle, Clone: SPM332, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23280810ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23280810

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Concentration 0.2mg/mL
Antigen Actin Alpha 2 Smooth Muscle
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P62736
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 59
Reconstitution Protein A purified
Clone SPM332
Conjugate Unlabeled
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep