Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 2 Smooth Muscle, Clone: SPM332, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476001

  • / Each
 

Description & Specifications

Specifications

Antigen Actin Alpha 2 Smooth Muscle
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM332
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P62736
Host Species Murine
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 59
Test Specificity This MAb is highly specific to actin from smooth muscles. Its epitope lies in the first four N-terminal amino acids.
Reconstitution Protein A purified
Target Species Baboon
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Feline
Target Species Goat
Target Species Guinea Pig
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Primate
Target Species Rabbit
Target Species Rat
Target Species Sheep