Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 2 Smooth Muscle, Clone: 1A4/asm-1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Actin Alpha 2 Smooth Muscle
Clone 1A4/asm-1
Concentration 1mg/mL
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Gene Accession No. P62736
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23452201

 
novus biologicals canada
NBP23452201MG
0.1mg Each for $384.00

NBP23452202

 
novus biologicals canada
NBP23452202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Actin Alpha 2 Smooth Muscle
Clone 1A4/asm-1
Concentration 1mg/mL
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Gene Accession No. P62736
Gene ID (Entrez) 59
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting