Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 2 Smooth Muscle, Clone: 1A4/asm-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452202

  • / Each
 

Description & Specifications

Specifications

Antigen Actin Alpha 2 Smooth Muscle
Gene ID (Entrez) 59
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Baboon
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Feline
Cross Reactivity Goat
Cross Reactivity Guinea Pig
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Cross Reactivity Sheep
Primary or Secondary Primary
Immunogen N-Terminal decapeptide of alpha smooth muscle isoform of actin and conjugated to KLH
Reconstitution Protein A purified
Concentration 1mg/mL
Host Species Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Isotype IgG2a, κ
Gene Accession No. P62736
Quantity 0.2mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Clone 1A4/asm-1
Format Purified