Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 1 Skeletal Muscle, Clone: 5C5.F8.C7, Novus Biologicals™

$307.00 - $473.61

Specifications

Antigen Actin Alpha 1 Skeletal Muscle
Clone 5C5.F8.C7
Concentration 0.2mg/mL
Immunogen N-terminal decapeptide of alpha skeletal muscle isoform of actin
Gene ID (Entrez) 58
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13526301

 
novus biologicals canada
NBP13526301ML
0.1mL Each for $307.00

NBP13526310

 
novus biologicals canada
NBP13526310ML
1mL Each for $473.61
Description & Specifications

Specifications

Antigen Actin Alpha 1 Skeletal Muscle
Clone 5C5.F8.C7
Concentration 0.2mg/mL
Immunogen N-terminal decapeptide of alpha skeletal muscle isoform of actin
Gene ID (Entrez) 58
Formulation 10mM PBS, pH 7.4
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgM
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)