Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 1 Skeletal Muscle, Clone: 5C5.F8.C7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13526301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13526301

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Conjugate Unlabeled
Quantity 0.1mL
Antigen Actin Alpha 1 Skeletal Muscle
Test Specificity This product is specific for Actin-Skeletal Muscle
Purification Method Purified
Primary or Secondary Primary
Formulation 10mM PBS, pH 7.4
Format Purified
Host Species Mouse
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 58
Reconstitution Protein A purified
Clone 5C5.F8.C7
Concentration 0.2mg/mL
Immunogen N-terminal decapeptide of alpha skeletal muscle isoform of actin
Regulatory Status RUO
Isotype IgM
Monoclonal or Polyclonal Monoclonal