Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 1 Skeletal Muscle, Clone: 5C5.F8.C7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13526370ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13526370

  • / Each
 

Description & Specifications

Specifications

Antigen Actin Alpha 1 Skeletal Muscle
Format Purified
Immunogen N-terminal decapeptide of alpha skeletal muscle isoform of actin
Isotype IgM
Storage Requirements Store at 4°C. Do not freeze.
Reconstitution Protein A purified
Formulation 10mM PBS, pH 7.4
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Purification Method Purified
Quantity 7mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 58
Test Specificity This product is specific for Actin-Skeletal Muscle
Clone 5C5.F8.C7
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human