Please login to your online account to display your discounted pricing

Mouse anti-Actin Alpha 1 Skeletal Muscle, Clone: 5C5.F8.C7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13526310ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13526310

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Format Purified
Formulation 10mM PBS, pH 7.4
Host Species Mouse
Quantity 1mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 58
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Antigen Actin Alpha 1 Skeletal Muscle
Immunogen N-terminal decapeptide of alpha skeletal muscle isoform of actin
Isotype IgM
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Test Specificity This product is specific for Actin-Skeletal Muscle
Clone 5C5.F8.C7
Concentration 0.2mg/mL
Purification Method Purified