Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: SPM333, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen ACTH
Clone SPM333
Concentration 0.2mg/mL
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Gene Accession No. P01189
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23291101

 
novus biologicals canada
NBP23291101ML
0.1mL Each for $139.00

NBP23291110

 
novus biologicals canada
NBP23291110ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen ACTH
Clone SPM333
Concentration 0.2mg/mL
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Gene Accession No. P01189
Gene ID (Entrez) 5443
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting