Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: SPM333, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23291110ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23291110

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Regulatory Status RUO
Gene ID (Entrez) 5443
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Reconstitution Protein A purified
Host Species Mouse
Gene Accession No. P01189
Antigen ACTH
Test Specificity Expected to show broad species reactivity.
Clone SPM333
Concentration 0.2mg/mL