Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: SPM333, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476101

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Gene ID (Entrez) 5443
Quantity 0.1mg
Isotype IgG1, κ
Primary or Secondary Primary
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Reconstitution Protein A purified
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Purification Method Purified
Clone SPM333
Antigen ACTH
Host Species Mouse
Concentration 1mg/mL
Test Specificity This MAb is specific to Synacthen (aa1-24 of ACTH); does not react with CLIP (aa17-39 of ACTH). Expected to show broad species reactivity.
Regulatory Status RUO
Gene Accession No. P01189