Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: SPM333, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476102

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01189
Host Species Mouse
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Primary or Secondary Primary
Gene ID (Entrez) 5443
Test Specificity This MAb is specific to Synacthen (aa1-24 of ACTH); does not react with CLIP (aa17-39 of ACTH). Expected to show broad species reactivity.
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled
Antigen ACTH
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone SPM333