Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: AH26 + 57, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen ACTH
Clone AH26 + 57
Concentration 0.2mg/mL
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH (AH26); N-terminal fragment of human ACTH conjugated to KLH (57)
Gene Accession No. P01189
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23428401

 
novus biologicals canada
NBP23428401ML
0.1mL Each for $139.00

NBP23428405

 
novus biologicals canada
NBP23428405ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen ACTH
Clone AH26 + 57
Concentration 0.2mg/mL
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH (AH26); N-terminal fragment of human ACTH conjugated to KLH (57)
Gene Accession No. P01189
Gene ID (Entrez) 5443
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting