missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: AH26 + 57, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen ACTH
Clone AH26 + 57
Concentration 1mg/mL
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH (AH26); N-terminal fragment of human ACTH conjugated to KLH (57)
Gene Accession No. P01189
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23455001 Novus Biologicals Canada
NBP23455001MG
0.1mg Each for $384.00
NBP23455002 Novus Biologicals Canada
NBP23455002MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

ACTH
1mg/mL
P01189
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
AH26 + 57
Synthetic peptide corresponding to aa1-24 of human ACTH (AH26); N-terminal fragment of human ACTH conjugated to KLH (57)
5443
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot