Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: 2F6, Novus Biologicals™

$321.00 - $362.00

Specifications

Antigen ACTH
Clone 2F6
Concentration 0.2mg/mL
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Gene ID (Entrez) 5443
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14216701

 
novus biologicals canada
NBP14216701ML
0.1mL Each for $362.00

NBP14216705

 
novus biologicals canada
NBP14216705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen ACTH
Clone 2F6
Concentration 0.2mg/mL
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Gene ID (Entrez) 5443
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting