Please login to your online account to display your discounted pricing

Mouse anti-ACTH, Clone: 2F6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14216701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14216701

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen ACTH
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Synthetic peptide corresponding to aa1-24 of human ACTH
Isotype IgG1
Gene ID (Entrez) 5443
Host Species Mouse
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 2F6
Conjugate Unlabeled
Reconstitution Protein A purified
Concentration 0.2mg/mL
Format Purified