Please login to your online account to display your discounted pricing

Mouse anti-ABCG2/CD338, Clone: 5D3, Novus Biologicals™

$278.00 - $354.00

Specifications

Antigen ABCG2/CD338
Clone 5D3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was ABCG2.
Gene ID (Entrez) 9429
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14316501

 
novus biologicals canada
NBP14316501MG
0.1mg Each for $354.00

NBP14316503

 
novus biologicals canada
NBP1431650025MG
0.025mg Each for $278.00
Description & Specifications

Specifications

Antigen ABCG2/CD338
Clone 5D3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was ABCG2.
Gene ID (Entrez) 9429
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry