Please login to your online account to display your discounted pricing

Mouse anti-ABCG2/CD338, Clone: 5D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14316501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14316501

  • / Each
 

Description & Specifications

Specifications

Antigen ABCG2/CD338
Applications Flow Cytometry
Clone 5D3
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Immunogen The immunogen for this antibody was ABCG2.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 9429