Please login to your online account to display your discounted pricing

Mouse anti-ABCG2/CD338, Clone: 5D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431650025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14316503

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Monoclonal or Polyclonal Monoclonal
Antigen ABCG2/CD338
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Isotype IgG2b, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Immunogen The immunogen for this antibody was ABCG2.
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 9429
Clone 5D3
Concentration 0.2mg/mL
Conjugate Unlabeled