Please login to your online account to display your discounted pricing

Mouse anti-6-Histidine Epitope Tag, Clone: 2A8, Novus Biologicals™

$307.00 - $362.00

Specifications

Antigen 6-Histidine Epitope Tag
Clone 2A8
Concentration 1mg/mL
Immunogen 6x His synthetic peptide coupled to KLH
Formulation PBS, pH 7.4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14128801

 
novus biologicals canada
NBP14128801ML
0.1mL Each for $307.00

NBP14128802

 
novus biologicals canada
NBP14128802ML
0.2mL Each for $362.00
Description & Specifications

Specifications

Antigen 6-Histidine Epitope Tag
Clone 2A8
Concentration 1mg/mL
Immunogen 6x His synthetic peptide coupled to KLH
Formulation PBS, pH 7.4
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Non-species specific
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting