Please login to your online account to display your discounted pricing

Mouse anti-6-Histidine Epitope Tag, Clone: 2A8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14128802ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14128802

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Non-species specific
Antigen 6-Histidine Epitope Tag
Formulation PBS, pH 7.4
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen 6x His synthetic peptide coupled to KLH
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Quantity 0.2mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity His-Tag (2A8) Mouse Monoclonal Antibody recognizes over-expressed or recombinant proteins containing the 6x His epitope tag.
Host Species Mouse
Clone 2A8
Concentration 1mg/mL