Please login to your online account to display your discounted pricing

Mouse anti-6-Histidine Epitope Tag, Clone: 2A8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14128801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14128801

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen 6x His synthetic peptide coupled to KLH
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen 6-Histidine Epitope Tag
Format Affinity Purified
Formulation PBS, pH 7.4
Isotype IgG1
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 2A8
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Non-species specific
Test Specificity His-Tag (2A8) Mouse Monoclonal Antibody recognizes over-expressed or recombinant proteins containing the 6x His epitope tag.