Please login to your online account to display your discounted pricing

Mouse anti-5'-Nucleotidase/CD73, Clone: AD2, APC, Novus Biologicals™

$192.00 - $386.00

Specifications

Antigen 5'-Nucleotidase/CD73
Clone AD2
Immunogen The immunogen for this antibody is CD73.
Gene ID (Entrez) 4907
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200297

 
novus biologicals canada
NBP200297100TESTS
100 Tests Each for $386.00

NBP20029725

 
novus biologicals canada
NBP20029725TESTS
25 Tests Each for $192.00
Description & Specifications

Specifications

Antigen 5'-Nucleotidase/CD73
Clone AD2
Immunogen The immunogen for this antibody is CD73.
Gene ID (Entrez) 4907
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry