Please login to your online account to display your discounted pricing

Mouse anti-5'-Nucleotidase/CD73, Clone: AD2, APC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP20029725TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20029725

  • / Each
 

Description & Specifications

Specifications

Antigen 5'-Nucleotidase/CD73
Applications Flow Cytometry
Clone AD2
Conjugate APC
Cross Reactivity Human
Format Affinity Purified
Host Species Mouse
Immunogen The immunogen for this antibody is CD73.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4907