Please login to your online account to display your discounted pricing

Mouse anti-5'-Nucleotidase/CD73, Clone: AD2, APC, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP200297100TESTS

 View all options for this product


For Research Use Only

Catalog No. NBP200297


Description & Specifications


Antigen 5'-Nucleotidase/CD73
Applications Flow Cytometry
Clone AD2
Conjugate APC
Cross Reactivity Human
Format Affinity Purified
Host Species Mouse
Immunogen The immunogen for this antibody is CD73.
Isotype IgG1, κ
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4907