Please login to your online account to display your discounted pricing
Mott Manufacturing Steel Casework Standing Height Base Cabinet, Full Door Units
Base cabinet with hinged doors and removable shelf
$431.00 - $670.46
Products
Catalog Number Mfr. No. Price Quantity    
36-104-1384
mott lab inc
1110011
Each for $431.00
36-104-1392
mott lab inc
1410011
Each for $627.88
36-104-1395
mott lab inc
1510011
Each for $670.46
Description & Specifications
  • Furniture grade cold rolled steel with high pre and post consumer recycled content
  • Laboratory grade powder coating - Warm Gray 601006
  • Contemporary inset design
  • Brushed aluminum bar handle(s) 10.1cm (4 in.) and Four levelling feet
  • Five-knuckle stainless steel hinges
  • Full depth single shelf 2.54cm (1 in.) thick, adjustable in 1.27cm (0.5 in.) increments
  • Full height removable back panel(s)
  • Quality doors include sound deadening material
Meets SEFA 8 performance standards