Please login to your online account to display your discounted pricing
Mott Manufacturing Steel Casework Specialty Cabinet, Insulated Solvent Units
Insulated base cabinet with one shelf for solvent storage
$1,282.37 - $1,933.32
Products
Catalog Number Mfr. No. Price Quantity    
36-104-1448
mott lab inc
1232011
Each for $1,282.37
36-104-1449
mott lab inc
1232111
Each for $1,282.37
36-104-1451
mott lab inc
1332011
Each for $1,729.35
36-104-1453
mott lab inc
1432011
Each for $1,800.30
36-104-1455
mott lab inc
1412011
Each for $1,837.54
36-104-1457
mott lab inc
1532011
Each for $1,933.32
Description & Specifications
  • Furniture grade cold rolled steel with high pre and post consumer recycled content
  • Laboratory grade powder coating - Warm Gray 601006
  • Contemporary inset design
  • Three point locking self latching handle
  • Continuous hinge(s) and Four levelling feet
  • Insulated double wall construction 5.08cm (2 in.)
  • Floor with 5.08cm (2 in.) liquid tight lip to contain spills
  • Meets National Fire Protection Association standards
Meets SEFA 8 performance standards